CHIRALPAK® IG 250X4.6 MM 5ΜM ANALYTICAL COLUMN 1 * 1 items
Catalog # DAIC87325
Supplier: CHIRAL TECHNOLOGIES
CHIRALPAK® IG 250X4.6 MM 5ΜM ANALYTICAL COLUMN 1 * 1 items
Catalog # DAIC87325
Supplier: CHIRAL TECHNOLOGIES
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
CHIRALPAK® IH 250 x 4,6 mm 5 µm Analytical Column 1 * 1 items
Supplier: CHIRAL TECHNOLOGIES
Immobilised polysaccharide columns
Supplier: CHIRAL TECHNOLOGIES
Protein based chiral HPLC columns, CHIRALPAK® AGP
Supplier: CHIRAL TECHNOLOGIES
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals