Order Entry
Italy
ContactUsLinkComponent
For quantitative detection of mouse Legumain(total) in cell culture supernates, cell lysates, tissue homogenates, serum and plasma ( heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1567
Supplier:  Boster Bio
undefined
For quantitative detection of mouse Legumain(total) in cell culture supernates, cell lysates, tissue homogenates, serum and plasma ( heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1567
Supplier:  Boster Bio