Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
295×197×18 mm
Catalog # SAMPA674-295
Supplier:   Sampling Systems
Instrument trays
Catalog # SAMPA674-295
Supplier:  Sampling Systems
Instrument trays
Instrument trays
295×197×18 mm
Price
42,40 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Made from 304 stainless steel.
  • Good chemical resistance, very good temperature resistance
  • Crevice free design
  • High quality polished finish
Ideal for general laboratory use as well as for carrying surgical instruments, sampling tools etc.
Documents
Specifications
  • Seq:
    02
  • L×P×A:
    295×197×18 mm