Order Entry
Italy
Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
215×160×18 mm
Catalog # SAMPA674-215
Supplier:   Sampling Systems
Instrument trays
Catalog # SAMPA674-215
Supplier:  Sampling Systems
Instrument trays
Instrument trays
215×160×18 mm
Price
27,90 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Made from 304 stainless steel.
  • Good chemical resistance, very good temperature resistance
  • Crevice free design
  • High quality polished finish
Ideal for general laboratory use as well as for carrying surgical instruments, sampling tools etc.
Documents
Specifications
  • Seq:
    01
  • W×D×H:
    215×160×18 mm