Order Entry
Italy
ContactUsLinkComponent
This product is prepared by labeling high quality goat anti-mouse IgG isotype-specific secondary antibody conjugates with biotin. Th e conjugates are prepared from affinity-purified antibodies that react with Fc portion of the heavy chain of mouse Ig 1 * 50 µl
Catalog # 20473-50UL
Supplier:  Biotium
This product is prepared by labeling high quality goat anti-mouse IgG isotype-specific secondary antibody conjugates with biotin. Th e conjugates are prepared from affinity-purified antibodies that react with Fc portion of the heavy chain of mouse Ig 1 * 50 µl
Catalog # 20473-50UL
Supplier:  Biotium
Frequently Bought Together