Order Entry
Italy
ContactUsLinkComponent
PERKINELMER OFFERS THE TITAN MPS MICROWAVE SAMPLE PREPARATION SYSTEMS, CONSUMABLES AND ACCESSORIES FOR YOUR LAB NEEDS. 8 POSITION TU RNTABLE SYSTEM WITH HIGH PRESSURE 100 ML (100 BAR) DIGESTION VESSELS (COMPLETE ASSEMBLY) FOR TITAN MPS 8 AND 16 POSIT 1 * 1 items
Catalog # PERCN3131011
Supplier:  PerkinElmer
PERKINELMER OFFERS THE TITAN MPS MICROWAVE SAMPLE PREPARATION SYSTEMS, CONSUMABLES AND ACCESSORIES FOR YOUR LAB NEEDS. 8 POSITION TU RNTABLE SYSTEM WITH HIGH PRESSURE 100 ML (100 BAR) DIGESTION VESSELS (COMPLETE ASSEMBLY) FOR TITAN MPS 8 AND 16 POSIT 1 * 1 items
Catalog # PERCN3131011
Supplier:  PerkinElmer
Frequently Bought Together