Order Entry
Italy
ContactUsLinkComponent
MATRIX: HYDROCARBON OIL, SIZE: 400 G. METALLO-ORGANIC METALS IN HYDROCARBON OIL. ACCURACY ENSURED BY QUALITY TESTING WITH NIST STAND ARD REFERENCE MATERIALS WHEN AVAILABLE. CERTIFICATE OF ANALYSIS SUPPLIED WITH EACH STANDARD. 1 * 1 items
Catalog # PERCN9308320
Supplier:  PerkinElmer
MATRIX: HYDROCARBON OIL, SIZE: 400 G. METALLO-ORGANIC METALS IN HYDROCARBON OIL. ACCURACY ENSURED BY QUALITY TESTING WITH NIST STAND ARD REFERENCE MATERIALS WHEN AVAILABLE. CERTIFICATE OF ANALYSIS SUPPLIED WITH EACH STANDARD. 1 * 1 items
Catalog # PERCN9308320
Supplier:  PerkinElmer