Order Entry
Italy
ContactUsLinkComponent
This product is prepared by labeling highly cross-adsorbed goat anti-rabbit IgG (H+L) with the green fluorescent dye CF™532. To mini mize cross-reactivity, the antibody has been adsorbed against human, mouse, and rat serum. 1 * 50 µl
Catalog # 20469-50UL
Supplier:  Biotium
This product is prepared by labeling highly cross-adsorbed goat anti-rabbit IgG (H+L) with the green fluorescent dye CF™532. To mini mize cross-reactivity, the antibody has been adsorbed against human, mouse, and rat serum. 1 * 50 µl
Catalog # 20469-50UL
Supplier:  Biotium
Frequently Bought Together