This product is prepared by labeling highly cross-adsorbed goat anti-mouse IgG (H+L) with the green fluorescent dye CF™532. To mini mize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, rabbit, and swine serum. 1 * 50 µl
Codice catalogo 20468-50UL
Supplier: Biotium
Frequently Bought Together