This product is prepared by single labeling (DOL=1) highly cross-adsorbed donkey anti-rabbit IgG (H+L) with near-IR fluorescent dye CF™750ST for (d)-STORM super-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed aga h 1 * 50 µl
Codice catalogo 20828-50UL
Supplier: Biotium
Frequently Bought Together