Order Entry
Italy
ContactUsLinkComponent
This product is prepared by single labeling (DOL=1) highly cross-adsorbed donkey anti-rabbit IgG (H+L) with CF™535ST for (d)-STORM s uper-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goa 1 * 50 µl
Codice catalogo 20824-50UL
Supplier:  Biotium
This product is prepared by single labeling (DOL=1) highly cross-adsorbed donkey anti-rabbit IgG (H+L) with CF™535ST for (d)-STORM s uper-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goa 1 * 50 µl
Codice catalogo 20824-50UL
Supplier:  Biotium
Frequently Bought Together