Lid for quartz inceneration crucible in application temperatures of up to 1050°C, transparent, high chemical resistance and purity. The durability or operating cycles of our quartz glass product mainly depends on applications. For application temper 1 * 1 items
Catalog # TSLA75701
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together