38 mm PP Closure for 125 mL Bottle. Process: Particulate Cleaning & Autoclave. Packaging: Single autoclave bag each cap. 72 per case. Certification: C of A Endotoxin (Report Actual Results), C of A USP Particles, C of A USP Sterility, Certificate of 1 * 72 items
Catalog # NALGCE-N6MM38AP-EP
Supplier: Thermo Fisher Scientific
Frequently Bought Together