Order Entry
Italy
ContactUsLinkComponent
BioSpectrum 815, MOTORISED fixed 25mm, f/0.95, MOTORISED Lift815 camera: 8.1 mp, 16 bit monochrome, digital camera with mount, with binning capabilities, cooling to -57C from ambient. MOTORISED 25mm, f/0.95 lens, close up diopter, USB2.0 connection. 1 * 1 items
Catalog # UVPA97-0714-02
BioSpectrum 815, MOTORISED fixed 25mm, f/0.95, MOTORISED Lift815 camera: 8.1 mp, 16 bit monochrome, digital camera with mount, with binning capabilities, cooling to -57C from ambient. MOTORISED 25mm, f/0.95 lens, close up diopter, USB2.0 connection. 1 * 1 items
Catalog # UVPA97-0714-02
Frequently Bought Together