Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Agarose, basic first

100 g
Catalog # ICNA11AGAF0100
Supplier:   MP Biomedicals
Agarose, basic first
Catalog # ICNA11AGAF0100
Supplier:  MP Biomedicals
CAS Number:  9012-36-6
Agarose, basic first
Agarose, basic first

100 g

CAS Number:  9012-36-6
Price
75,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Basic agarose premier is ideal for routine rapid separation of DNA and RNA fragments as well as PCR products, the preparation of plasmids, and for screening, cloning and blotting techniques.

Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.

  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pk:
    100 g

Specifications