Order Entry
Italy
ContactUsLinkComponent
Wash solution included in the FastDNA and all FastDNA Spin Kits Used to remove impurities once the DNA is bound to the Binding Matrix 1 * 12 mL
Catalog # ICNA116540405
Supplier:  MP Biomedicals
Wash solution included in the FastDNA and all FastDNA Spin Kits Used to remove impurities once the DNA is bound to the Binding Matrix 1 * 12 mL
Catalog # ICNA116540405
Supplier:  MP Biomedicals
Frequently Bought Together