Order Entry
Italy
ContactUsLinkComponent
DIATOMACEOUS EARTH ACID-WASHED. Highly insoluble, inert material for use as a filtration aid.Acid-washedPurity: 95% SiO2Suitable fo r most routine filtrations.Light pink to tan powder. 1 * 100 g
Catalog # ICNA0215760780
Nome del fornitore:  MP Biomedicals
DIATOMACEOUS EARTH ACID-WASHED. Highly insoluble, inert material for use as a filtration aid.Acid-washedPurity: 95% SiO2Suitable fo r most routine filtrations.Light pink to tan powder. 1 * 100 g
Catalog # ICNA0215760780
Nome del fornitore:  MP Biomedicals
Frequently Bought Together