Order Entry
Italy
ContactUsLinkComponent
ATRIAL NATRIURETIC PEPTIDE, Fragment 3-28. (ANP Fragment 8-33 ANP Fragment 125-150)Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Gly-Ala- Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-TyrSpecies: RatPurity: 98% 1 * 100 µG
Catalog # ICNA0215989080
Nome del fornitore:  MP Biomedicals
ATRIAL NATRIURETIC PEPTIDE, Fragment 3-28. (ANP Fragment 8-33 ANP Fragment 125-150)Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Gly-Ala- Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-TyrSpecies: RatPurity: 98% 1 * 100 µG
Catalog # ICNA0215989080
Nome del fornitore:  MP Biomedicals
Frequently Bought Together