Order Entry
Italy
Orders LinkContactUsLinkComponent
Paraformaldehyde, powder

Paraformaldehyde, powder

500 g
Catalog # ICNA02150146.5
Supplier:   MP Biomedicals
Paraformaldehyde, powder
Catalog # ICNA02150146.5
Supplier:  MP Biomedicals
CAS Number:  30525-89-4
Paraformaldehyde, powder

500 g

CAS Number:  30525-89-4
Price
86,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Soluble in water (hot water - ~37% w/v; slowly soluble in cold water). Aqueous solutions will develop formaldehyde over time. Insoluble in alcohol or ether.

Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • UN:2213
  • ADR:4.1,III
Specifications

Specifications

  • Pk:
    500 g

Specifications