Order Entry
Italy
ContactUsLinkComponent
From Streptomyces sp.Antibiotic Inhibits many biochemical functions such as synthesis of glycoproteins, procollagen and peptidoglycans and IgA and IgE secretion by plasma cells. Also affects the fibroblast interferon-cell system. 1 * 10 mg
Codice catalogo ICNA0215002810
Supplier:  MP Biomedicals
From Streptomyces sp.Antibiotic Inhibits many biochemical functions such as synthesis of glycoproteins, procollagen and peptidoglycans and IgA and IgE secretion by plasma cells. Also affects the fibroblast interferon-cell system. 1 * 10 mg
Codice catalogo ICNA0215002810
Supplier:  MP Biomedicals
Frequently Bought Together