Order Entry
Italy
ContactUsLinkComponent
DEOXY-BIGCHAP. [N,N'-Bis(3-D-gluconamidopropyl)- deoxy-cholamide] New analog of CHAPS and CHAPSO with less electrostatic interaction s, making it excellent for use in anion exchange chromatography. 1 * 100 mg
Catalog # ICNA02152291.1
Supplier:  MP Biomedicals
DEOXY-BIGCHAP. [N,N'-Bis(3-D-gluconamidopropyl)- deoxy-cholamide] New analog of CHAPS and CHAPSO with less electrostatic interaction s, making it excellent for use in anion exchange chromatography. 1 * 100 mg
Catalog # ICNA02152291.1
Supplier:  MP Biomedicals
Frequently Bought Together