Order Entry
Italy
ContactUsLinkComponent
From Porcine Pancreas E.C.3.1.1.3 Purified and standardized. Activity: 16,000 units/gmUnit Definition: One unit will liberate 10 0 µ moles fatty acid per hour at pH 7.8 and 37°C 1 * 1 g
Catalog # ICNA0210081701
Supplier:  MP Biomedicals
From Porcine Pancreas E.C.3.1.1.3 Purified and standardized. Activity: 16,000 units/gmUnit Definition: One unit will liberate 10 0 µ moles fatty acid per hour at pH 7.8 and 37°C 1 * 1 g
Catalog # ICNA0210081701
Supplier:  MP Biomedicals
Frequently Bought Together