FERRIC NITRATE NONAHYDRATE ACS REAGENT GRADE. Purity: >98% 1 * 100 g
ICNA0219404680
FERRIC NITRATE NONAHYDRATE ACS REAGENT GRADE. Purity: >98% 1 * 100 g
ICNA0219404680
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Toluene ≥98%, TECHNICAL
: VWR Chemicals