Order Entry
Italy
ContactUsLinkComponent
Progesterone, cyclodextrin-encapsulated.Water Soluble.Approximately 70 mg progesterone per gram using a balance consisting of 2-hydroxypropyl- -cyclodextrin. Typically used in cell culture applications. This is a suspected carcinogen. 1 * 100 mg
Catalog # ICNA0219935280
Supplier:  MP Biomedicals
Progesterone, cyclodextrin-encapsulated.Water Soluble.Approximately 70 mg progesterone per gram using a balance consisting of 2-hydroxypropyl- -cyclodextrin. Typically used in cell culture applications. This is a suspected carcinogen. 1 * 100 mg
Catalog # ICNA0219935280
Supplier:  MP Biomedicals
Frequently Bought Together