Order Entry
Italy
ContactUsLinkComponent
From Aspergillus niger Lyophilized Activity : 3-9 units per mg protein.Unit Definition : One unit will liberate 1.0 µmole of galacturonic acid per min at pH 4.0 at 25°C. 1 * 25 KU
Catalog # ICNA0219897925
Supplier:  MP Biomedicals
From Aspergillus niger Lyophilized Activity : 3-9 units per mg protein.Unit Definition : One unit will liberate 1.0 µmole of galacturonic acid per min at pH 4.0 at 25°C. 1 * 25 KU
Catalog # ICNA0219897925
Supplier:  MP Biomedicals
Frequently Bought Together