Order Entry
Italy
ContactUsLinkComponent
Deionized water treated with 0.001% diethylpyrocarbonate (depc). filtered through 0.2 micron filter and autoclaved to yield a sterile solution completely free of detectable dnase (both exo and endo) and rnase. 1 * 500 mL
  ICNA04821932
Deionized water treated with 0.001% diethylpyrocarbonate (depc). filtered through 0.2 micron filter and autoclaved to yield a sterile solution completely free of detectable dnase (both exo and endo) and rnase. 1 * 500 mL
  ICNA04821932
Frequently Bought Together