Cell standard rectangular with lid matched 10mm pathlength 10mm ID 2000µl working volume 12,5x45mm external dimensions 1 * 2 items
Catalog # SCLI80-2099-89
Supplier: BIOCHROM
Cell standard rectangular with lid matched 10mm pathlength 10mm ID 2000µl working volume 12,5x45mm external dimensions 1 * 2 items
Catalog # SCLI80-2099-89
Supplier: BIOCHROM