Order Entry
Italy
ContactUsLinkComponent
A reversible inhibitor of the auto and transphosphorylation kinase activity of the erbB receptor tyrosine kinase superfamily: EGFR ( IC50 = 8 µM), erbB-2 (IC50 = 49 nM), erbB3, and erbB4 (IC50 = 52 nM) 1 * 10 mg
Catalog # CAYM13329-10
Supplier:  Cayman Chemical
A reversible inhibitor of the auto and transphosphorylation kinase activity of the erbB receptor tyrosine kinase superfamily: EGFR ( IC50 = 8 µM), erbB-2 (IC50 = 49 nM), erbB3, and erbB4 (IC50 = 52 nM) 1 * 10 mg
Catalog # CAYM13329-10
Supplier:  Cayman Chemical
Frequently Bought Together