Order Entry
Italy
ContactUsLinkComponent
A GSK3 inhibitor (Ki = 4.9 nM) that crosses the blood brain barrier and inhibits tau phosphorylation (IC50 = 76 nM in vitro) 1 * 10 mg
Catalog # CAYM16728-10
Supplier:  Cayman Chemical
A GSK3 inhibitor (Ki = 4.9 nM) that crosses the blood brain barrier and inhibits tau phosphorylation (IC50 = 76 nM in vitro) 1 * 10 mg
Catalog # CAYM16728-10
Supplier:  Cayman Chemical
Frequently Bought Together