A potent, selective, brain permeable inhibitor of GSK3 and GSK3 (Kis = 6.9 and 31 nM, respectively) 1 * 25 mg
CAYM16676-25
A potent, selective, brain permeable inhibitor of GSK3 and GSK3 (Kis = 6.9 and 31 nM, respectively) 1 * 25 mg
CAYM16676-25
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection