Order Entry
Italy
ContactUsLinkComponent
A toxic antimetabolite for the tryptophan pathway in yeast that can be used to counterselect for TRP1, a commonly used genetic marke r in S. cerevisiae 1 * 1 g
Catalog # CAYM16669-1
Supplier:  Cayman Chemical
A toxic antimetabolite for the tryptophan pathway in yeast that can be used to counterselect for TRP1, a commonly used genetic marke r in S. cerevisiae 1 * 1 g
Catalog # CAYM16669-1
Supplier:  Cayman Chemical
Frequently Bought Together