Order Entry
Italy
ContactUsLinkComponent
An SGK1 inhibitor (IC50s = 62 and 103 nM for SGK1 and SGK2, respectively) that demonstrates greater than 30-fold selectivity for SGK 1 over Akt and other related kinases 1 * 5 mg
  CAYM17001-5
An SGK1 inhibitor (IC50s = 62 and 103 nM for SGK1 and SGK2, respectively) that demonstrates greater than 30-fold selectivity for SGK 1 over Akt and other related kinases 1 * 5 mg
  CAYM17001-5
Frequently Bought Together