Order Entry
Italy
ContactUsLinkComponent
An SGK1 inhibitor (IC50s = 62 and 103 nM for SGK1 and SGK2, respectively) that demonstrates greater than 30-fold selectivity for SGK 1 over Akt and other related kinases 1 * 25 mg
Catalog # CAYM17001-25
Supplier:  Cayman Chemical
An SGK1 inhibitor (IC50s = 62 and 103 nM for SGK1 and SGK2, respectively) that demonstrates greater than 30-fold selectivity for SGK 1 over Akt and other related kinases 1 * 25 mg
Catalog # CAYM17001-25
Supplier:  Cayman Chemical
Frequently Bought Together