Order Entry
Italy
ContactUsLinkComponent
A non-hydrolyzable phosphorothioate analog of cAMP that competitively inhibits cAMP-dependent protein kinases (PKA) I and II (IC50s = 12.5 and 4.5 µM, respectively) 1 * 500 µG
Catalog # CAYM16985-500
Supplier:  Cayman Chemical
A non-hydrolyzable phosphorothioate analog of cAMP that competitively inhibits cAMP-dependent protein kinases (PKA) I and II (IC50s = 12.5 and 4.5 µM, respectively) 1 * 500 µG
Catalog # CAYM16985-500
Supplier:  Cayman Chemical
Frequently Bought Together