A dual inhibitor of IGF1R kinase and IR (IC50s = 100 and 73 nM, respectively) 1 * 5 mg
CAYM14666-5
A dual inhibitor of IGF1R kinase and IR (IC50s = 100 and 73 nM, respectively) 1 * 5 mg
CAYM14666-5
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection