Order Entry
Italy
ContactUsLinkComponent
A serotonin 5-HT1B and 5-HT1D receptor agonist (IC50s = 9.3 and 7.3 nM, respectively) that also has affinity for the 5-HT1F receptor (IC50 = 17.8 nM) 1 * 5 mg
  CAYM14600-5
A serotonin 5-HT1B and 5-HT1D receptor agonist (IC50s = 9.3 and 7.3 nM, respectively) that also has affinity for the 5-HT1F receptor (IC50 = 17.8 nM) 1 * 5 mg
  CAYM14600-5
Frequently Bought Together