β-Alanine
Fiber drum, 50 kg
Catalog # SPCMAL120-50KGBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
- CAS Number:107-95-9
- Formula:C₃H₇NO₂
- MDL Number:MFCD00008200
- Molecular Weight:89.09 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Fiber drum
- Pk:50 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



