Order Entry
Italy
Orders LinkContactUsLinkComponent
 

β-Alanine

Poly bottle, 2,5 kg
Catalog # SPCMAL120-2.5KG
Supplier:   Spectrum Chemical
β-Alanine
Catalog # SPCMAL120-2.5KG
Supplier:  Spectrum Chemical
CAS Number:  107-95-9
β-Alanine

Poly bottle, 2,5 kg

CAS Number:  107-95-9
Price
700,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    2,5 kg

Specifications