Polysorbate 40 NF
200 L
Catalog # SPCMPO134-200LTBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
- CAS Number:9005-66-7
- Formula:C₆₂H₁₂₂O₂₆
- MDL Number:MFCD00165345
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:200 L



