Order Entry
Italy
Orders LinkContactUsLinkComponent
Hydroxyurea ≥97% USP
12 kg
  SPCMH1181-12KGBL
Hydroxyurea ≥97% USP
  SPCMH1181-12KGBL
 :  127-07-1
Hydroxyurea ≥97% USP
12 kg
 :  127-07-1
13 500,00 €
1 * 12 kg
Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
 :  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  •  :127-07-1
  •  :CH₄N₂O₂
  •  :MFCD00007943
  •  :76.05 g/mol
  •  :13,04874
  •  :296 °C (1013 hPa)
  •  :142…144 °C
  •  :Ambient
  •  :204-821-7
  • Assay:
    97% Min.
  • Pk:
    12 kg