Oxidizing Reagent B, 0,02M Iodine In Thf : Pyridine : Water (69 22 11(W/W/W) Novabiochem 1 * 450 mL
8.57013.0450
Oxidizing Reagent B, 0,02M Iodine In Thf : Pyridine : Water (69 22 11(W/W/W) Novabiochem 1 * 450 mL
8.57013.0450
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection