Order Entry
Italy
ContactUsLinkComponent
Mixture VOC 27 10 x 1,2 mL VOC Mix 26 je 5000 ìg/mL in Methanol Benzol / Toluol o-Xylol / p- Xylol Ethylbenzol / Styrol Trichlorethylen / Tetrachlorethylen Naphthalin / 2- Ethylhexanol 2,6-Dimethylphenol / Dicyclohexylamin Phenol / Di-n-butylphthalat 1 * 1 KIT
Catalog # PROOSL20507-KIT
Mixture VOC 27 10 x 1,2 mL VOC Mix 26 je 5000 ìg/mL in Methanol Benzol / Toluol o-Xylol / p- Xylol Ethylbenzol / Styrol Trichlorethylen / Tetrachlorethylen Naphthalin / 2- Ethylhexanol 2,6-Dimethylphenol / Dicyclohexylamin Phenol / Di-n-butylphthalat 1 * 1 KIT
Catalog # PROOSL20507-KIT