Order Entry
Italy
ContactUsLinkComponent
Llama Ig Fraction contains total llama Ig from normal serum, e.g. serum of non immunized animals and is excellent for use as blockin g reagent in immunoassays. ,Llama Ig Fraction contains total llama Ig from normal serum, e.g. serum of non immunized 1 * 10 mg
Catalog # AGRIAS152842
Supplier:  Agrisera
Llama Ig Fraction contains total llama Ig from normal serum, e.g. serum of non immunized animals and is excellent for use as blockin g reagent in immunoassays. ,Llama Ig Fraction contains total llama Ig from normal serum, e.g. serum of non immunized 1 * 10 mg
Catalog # AGRIAS152842
Supplier:  Agrisera