Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
AMMONIUM ION CHROMATOGRAPHY STANDARD IN H2O,AMMONIUM - NH4(+) @100µG/ML 1 * 100 mL
Catalog # PROOVHGINH4100-100
Supplier:
LGC Standards PROMOCHEM
Expand image
AMMONIUM ION CHROMATOGRAPHY STANDARD IN H2O,AMMONIUM - NH4(+) @100µG/ML 1 * 100 mL
Catalog # PROOVHGINH4100-100
Supplier:
LGC Standards PROMOCHEM
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Calcium carbonate, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
pH buffer solutions
Supplier: Hach