GLYCOLATE ION CHROMATOGRAPHY STANDARD IN H2O,GLYCOLATE - C2H3O3(-) @ 1,000UG/ML 1 * 100 mL
PROOVHG-IGLY-100
GLYCOLATE ION CHROMATOGRAPHY STANDARD IN H2O,GLYCOLATE - C2H3O3(-) @ 1,000UG/ML 1 * 100 mL
PROOVHG-IGLY-100
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection