Order Entry
Italy
ContactUsLinkComponent
50µG/G AG, AL, B, BA, CA, CD, CR, CU, FE, K, MG, MN, MO, NA, NI, P, PB, SB, SI, SN, TI, V, ZN IN HYDROCARBON OIL 1 * 1 items
  PROOG-V23-50-200G
50µG/G AG, AL, B, BA, CA, CD, CR, CU, FE, K, MG, MN, MO, NA, NI, P, PB, SB, SI, SN, TI, V, ZN IN HYDROCARBON OIL 1 * 1 items
  PROOG-V23-50-200G
Frequently Bought Together