STRONTIUM IN 2% HNO3, SR: 100 µG/ML NOM., NATURAL ABUNDANCE VALUES 1 * 50 mL
PROOVHG-LISSR-50
STRONTIUM IN 2% HNO3, SR: 100 µG/ML NOM., NATURAL ABUNDANCE VALUES 1 * 50 mL
PROOVHG-LISSR-50
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection