JWH-210 ((4-ETHYLNAPHTHALEN-1-YL)(1-PENTYLINDOL-3-YL)METHANONE) 1.0 MG/ML IN ACETONITRILE 1 * 1 mL
Catalog # PROOAMP1275.67-11
Supplier: LGC Standards PROMOCHEM
JWH-210 ((4-ETHYLNAPHTHALEN-1-YL)(1-PENTYLINDOL-3-YL)METHANONE) 1.0 MG/ML IN ACETONITRILE 1 * 1 mL
Catalog # PROOAMP1275.67-11
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
CONDUCTIVITY STANDARD 1.3µS/CM 1 * 250 mL
Supplier: HAMILTON BONADUZ
pH buffer solutions, NIST/DIN
Supplier: Mettler - Toledo
Sucrose, Reference Standard
Supplier: USP
VWR®, Viscosity standards
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT