Order Entry
Italy
ContactUsLinkComponent
Rotavapor R-300 no bath, SJ24/40, S, P+G, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o SJ 24/40 Glass set: Vertical ref lux (S) P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300202S151
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ24/40, S, P+G, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o SJ 24/40 Glass set: Vertical ref lux (S) P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300202S151
Supplier:  BUCHI
Frequently Bought Together