Rotavapor R-300 B-305, SJ24/40, V, P+G, I-300P, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: Vertical (V) P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300152V151
Supplier: BUCHI
Frequently Bought Together